Laminar flow workstations, P range

Supplier: BASSAIRE
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P2HB P2HF P2VB P2VF P3HB P3HF P3VB P3VF P4HB P4HF P4VB P4VF P5HB P5HF P5VB P5VF P6HB P6HF P6VB P6VF
135-0026EA 0 GBP
135-0026 135-0027 135-0028 135-0029 135-0030 135-0031 135-0032 135-0033 135-0034 135-0035 135-0036 135-0037 135-0038 135-0039 135-0040 135-0041 135-0042 135-0025 135-0043 135-0044
Laminar flow workstations, P range
Fume Hoods and Enclosures Laminar Flow Cabinets
These laminar flow workstations have been designed to protect product and operating staff by maintaining a dust free ultra clean environment. The P-range is constructed in white powder coated high quality carbon steel with a 304 L stainless steel post formed worktop.

  • Combine safety and reliability
  • User-friendly and easy to clean and sanitize
  • Provide an aesthetic appearance
  • Choice of five sizes in horizontal (H) or vertical (V) flow configuration
  • Bench (B) or free standing (F) models
  • 13 amp socket outlet and variable speed controllers

A variety ofMore Product Information
Order Now

SPECIFICATIONS

Model P2H P2V P2V P3H P3H P3V P3V P4H P4V P5H P5V P5V P6H P6V
Environmentally Preferable
Fans Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected Double inlet, centrifugal, super silent and thermally protected
Air velocity 0,15 m/s* 0,15 m/s* 0,15 m/s* 0,23 m/s* 0,23 m/s* 0,23 m/s* 0,23 m/s* 0,31 m/s* 0,31 m/s* 0,39 m/s* 0,39 m/s* 0,39 m/s* 0,47 m/s* 0,47 m/s*
Lighting 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface 1000 Lux minimum at work surface
Noise level Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A) Level ISO 14644 at normal ambient levels of 35 − 40 dB(A)
Operating resistance Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa Maximum 230 Pa
Filters Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Pre-filter: 92% efficient at 5,0 micron
HEPA filter: 99,995% efficient at the MMPS (EN 1822 Class H 14)
Electrical supply 230− 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230− 240 V, 50/60 Hz single phase 230− 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230− 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230− 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase 230 - 240 V, 50/60 Hz single phase
Ext. W×D×H 680×720×1145 mm 660×660×1286 mm 660×660×1286 mm 985×720×1145 mm 985×720×1145 mm 660×965×1286 mm 660×965×1286 mm 1290×720×1145 mm 1270×660×1286 mm 1595×720×1145 mm 1270×660×1286 mm 1575×660×1286 mm 1900×720×1145 mm 1880×660×1286 mm
Int. W×D×H 580×505×579 mm 610×635×600 mm 610×635×600 mm 885×505×579 mm 885×505×579 mm 915×635×600 mm 915×635×600 mm 1190×505×579 mm 1220×635×600 mm 1495×505×579 mm 1525×635×600 mm 1525×635×600 mm 1800×505×579 mm 1830×635×600 mm
Ext.Ø×H 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm 720×1145 mm
Int.Ø×H 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm 505×579 mm
Weight 100 kg 85 kg 85 kg 125 kg 125 kg 100 kg 100 kg 150 kg 120 kg 189 kg 196 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR