Holmium tribromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

13203.09 13203.18
13203.09EA 59.1 GBP
13203.09 13203.18
Holmium tribromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Holmium tribromide hydrate

Purity: 99.99% CAS No.: 223911-98-6 Molecular Formula: HoBr3.xH2O Formula Weight: 404.66(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,4727 MDL No.: MFCD00209652

Formula: Br₃Ho·H₂O
Melting Pt: 914 °C
Storage Temperature: Ambient
MDL Number: MFCD00209652
CAS Number: 223911-98-6
EINECS: 237-524-6
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR