VWR®, ESD Chairs and Stools for Cleanrooms
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
132-1370EA 557 GBP
132-1370 132-1371 132-1372 132-1368 132-1369 132-1373
VWR®, ESD Chairs and Stools for Cleanrooms
Chairs
Ergonomic, auto-adaptive chairs or stools made from PU, for cleanrooms.
  • Auto-adaptive to users and to their positions
  • Facilitates an upright posture
  • Minimises spinal stress
  • Abrasion resistant
  • Black steel, five branch, scuff resistant, reinforced base
  • Black gas-lift column, with height adjustment and shock absorber
The innovative seat design consists of a short seat, tilted forward, split into two parts, based on blue silent blocks.
Special conductive PU connects to the metal parts of the chairs and stools, allowing electricity to be dischargedMore Product Information