Drum sumps and pallets
Supplier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18073 18074 18075 18076 18077 18078 18447 18449 31860 31861 31862 40046 40030
132-0557EA 191 GBP
132-0557 132-0558 132-0559 132-0560 132-0561 132-0562 132-0554 132-0556 MSGA31860 MSGA31861 MSGA31862 MSGA40046 MSGA40030
Drum sumps and pallets
Drum Storage
This sump pallets are made of PE with varied capacities from 60 to 400 L.
  • Compact and robust design
  • Approved for the safe storage of water pollutants
  • Swivel castors and handle for mobility
  • Blue in colour
Sump pallets with PE grid or galvanised grid are helps in direct placing on the floor and provide resistant against oil, acid and alkali.
Feet present with sumps are ideal for in-house transport of the empty sump pallet.