Nitronium hexafluorophosphate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

13151.06 13151.14
13151.06EA 94 GBP
13151.06 13151.14
Nitronium hexafluorophosphate
Nitronium hexafluorophosphate

CAS No.: 19200-21-6 Molecular Formula: NO2PF6 Formula Weight: 190.97 Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00031520

Formula: F₆NO₂P
Storage Temperature: Ambient
MDL Number: MFCD00031520
CAS Number: 19200-21-6
EINECS: 242-872-7
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR