Calcium chloride 89, pellets, purified, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

1313-07EA 664 GBP
1313-07 1313-01 1313-05
Calcium chloride 89, pellets, purified, J.T.Baker®
Calcium chloride

Pellets. Desiccant, 4 to 8 mesh.

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥89%
Assay (CaCl₂) ≥89 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.02 %
Trace Impurities - Arsenic (As) ≤1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR