Chemistry vacuum pumps and systems with valve control, dual application
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20733152 20733252 20733352 20737152 20737252 20737352
131-0022EA 4820 GBP
131-0022 131-0025 131-0028 131-0039 131-0042 131-0045
Chemistry vacuum pumps and systems with valve control, dual application
Pumps Vacuum Diaphragm Pumps
These chemistry pumping units use chemical resistant solenoid valves to maintain process control. Once the first boiling point is detected by the controller, it maintains the pressure at this level. Pumping units can be configured to run one or two independent processes at the same time.
  • VACUU·SELECT vacuum controller with graphical user interface simplifies work in the laboratory
  • Predefined applications save time and give reproducible results
  • Easy-to-use application editor allows own process sequences
  • Excellent environmental friendliness due to efficient
More Product Information