Propidium iodide

Supplier: Tonbo Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13-6990-T500 13-6990-T200
13-6990-T500EA 61.9 GBP
13-6990-T500 13-6990-T200
Propidium iodide
Propidium iodide

Propidium Iodide (PI) is a membrane-impermeant DNA binding dye that cannot penetrate viable cells. PI rapidly enters cells with compromised membranes and intercalates between base pairs allowing exclusion of non-viable cells from analysis of flow cytometry data. PI has a broad emission spectrum from 535-617 nm and can be detected in either FL2 or FL3 detectors. When used with Annexin V FITC, it is recommended to analyze PI in FL2.


  • Formulation: 10 mM NaH₂PO₄, 150 mM NaCl, 0,09% NaN₃, pH 7,2

Formula: C₂₇H₃₄I₂N₄
MDL Number: MFCD00011921
CAS Number: 25535-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR