VWR® Autoclavable Biohazard Bags, Polypropylene, Orange

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

129-4026EA 137 GBP
129-4026 129-4034 129-4042
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Bags Waste Disposal Bags

Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste or hazardous waste that must first be autoclaved.



  • Autoclavable up to 135 °C maximum

  • 200 mm twist ties included allows bag to expand and vent steam during cooling
  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Bag is printed with the universal biohazard symbol
  • Bag has temperature indicator, text 'Contents Have Been Autoclaved' turns dark to indicates successful autoclaving
  • Highly resistant to tears and punctures


Can also be

More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR