VWR® Maximum, Protection Covers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
129-1216EA 87.2 GBP
129-1216 129-1220
VWR® Maximum, Protection Covers
Covers
Manufactured from fluid-impervious material.
  • Full protection from fluids and light chemical splashes
  • Lowest level of particle shedding
  • Clean, low linting fabric
  • Tunnelled elastic, along the bottom perimeter, ensures a secure fit
These covers provide barrier protection against fluids, biologicals, particulates and light chemical splashes.
Bacterial Filtration Efficiency: 99,9%
Level I per Helmke Drum test
Certifications: Covers are rigorously tested and manufactured in an ISO certified facility, under stringent process controls, to ensure thatMore Product Information