VWR®, Zip-Seal Bags
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
129-0320EA 209 GBP
129-0320 129-0321 129-0322 129-0323 129-0324 129-0325 129-0293 129-0294 129-0295 129-0296 129-0297 129-0298 129-0299 129-0306 129-0307 129-0308 129-0309 129-0310 129-0311 129-0312 129-0313 129-0314 129-0315 129-0316 129-0317 129-0318 129-0319
VWR®, Zip-Seal Bags
Bags Sample Bags
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
  • Quick sealing with guide tracks on the pressure seal
  • Transparent PE for easy identification of the contents
  • Can be resealed repeatedly, flexible and tear-proof
Packaging: 100/bag, 10 bags/case.