Rubidium fluoride 99.975% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

12873.04 12873.09
12873.04EA 74.8 GBP
12873.04 12873.09
Rubidium fluoride 99.975% (metals basis), Puratronic®
Rubidium fluoride

Purity: 99.975% CAS No.: 13446-74-7 Molecular Formula: FRb Formula Weight: 104.47 Physical Form: Crystalline Density: 3.557 Refractive Index: 1.398 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 795° UN No.: 3288 MDL No.: MFCD00011191

Formula: FRb
Boiling Pt: 1408 °C (1013 hPa)
Melting Pt: 795 °C
Density: 3.557
Storage Temperature: Ambient
MDL Number: MFCD00011191
CAS Number: 13446-74-7
EINECS: 236-603-2
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR