Strontium sulphate ≥99.9965% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12870.06 12870.14
12870.06EA 53.1 GBP
12870.06 12870.14
Strontium sulphate ≥99.9965% (metals basis), Puratronic®
Strontium sulphate

Purity: 99.9965% CAS No.: 7759-02-6 Molecular Formula: O4SSr Formula Weight: 183.68 Physical Form: Powder Density: 3.960 Appearance: White Odour: Odourless Melting Point: 1600° Merck Reference: 14,8848 MDL No.: MFCD00036274

Formula: O₄SSr
Melting Pt: 1600 °C
Density: 3.960
Storage Temperature: Ambient
MDL Number: MFCD00036274
CAS Number: 7759-02-6
EINECS: 231-850-2
Merck Index: 14,8848

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR