Thallium ≥99.999% (metals basis), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

12714.0Y 12714.DO
12714.0YEA 1619 GBP
12714.0Y 12714.DO
Thallium ≥99.999% (metals basis), rods, Ø 12.7 mm (0.5 in)
Thallium

Purity: 99.999% CAS No.: 7440-28-0 Physical Form: ≈225g/150mm Odour: Odourless MDL No.: MFCD00134063

Formula: Tl
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 301 °C
Density: 11.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134063
CAS Number: 7440-28-0
EINECS: 231-138-1
UN: 1707
ADR: 6.1,II
Merck Index: 12,09391

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR