Anti-ALB Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

126705-1
126705-1EA 168 GBP
126705-1
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR