Potassium hexacyanoferrate(II) trihydrate ≥99%, Purum, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12639-25KG
12639-25KGEA 721 GBP
12639-25KG
Potassium hexacyanoferrate(II) trihydrate ≥99%, Purum, Fluka™
Potassium hexacyanoferrate (II) trihydrate

  • ≥95% for general laboratory use.

Formula: K₄Fe(CN)₆·3H₂O
Melting Pt: 70 °C (Elimination of water of crystallisation)
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2

Order Now

Specification Test Results

Assay Min. 99 %
Water insoluble matter Max. 0.02 %
Chloride (Cl) Max. 0.5 %
Sulfate (SO₄) Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR