Aluminium acetylacetonate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

12542.22 12542.36
12542.22EA 33 GBP
12542.22 12542.36
Aluminium acetylacetonate
Aluminium acetylacetonate

CAS No.: 13963-57-0 Molecular Formula: C15H21AlO6 Formula Weight: 324.31 Physical Form: Powder Density: 1.27 Melting Point: 192-194° Boiling Point: 314-316° Beilstein Reference: 4157942 UN No.: 3467 MDL No.: MFCD00000013

Formula: C₁₅H₂₁AlO₆
Boiling Pt: 314…316 °C
Melting Pt: 192…194 °C
Density: 1.27
Storage Temperature: Ambient
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR