VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
124802MEA 83.5 GBP
124802M
VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
Standards Organic Standards
17 components.
20 µg/ml : Aldrin; a-BHC; b-BHC; d-BHC; g-BHC; p,p'-DDD; p,p'-DDE; Dieldrin; Endosulfan I; Endrin aldehyde; Heptachlor; Heptachlor epoxide (Isomer B); Methoxychlor
100 µg/ml : p,p'-DDT; Endosulfan II; Endosulfan sulfate; Endrin