Pesticides Mix 2, EPA 608
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
124782CEA 50.6 GBP
124782C
Pesticides Mix 2, EPA 608
Standards Organic Standards
Chlordane (0,02 mg/ml) and Toxaphene (0,20 mg/ml) in methanol.