Perchloric acid 67-72%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

12415-250ML 12415-1L 12415-500ML
12415-250MLEA 616 GBP
12415-250ML 12415-1L 12415-500ML
Perchloric acid 67-72%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Perchloric acid

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: HClO₄
Boiling Pt: 203 °C
Melting Pt: -18 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 67 - 72 %
Silver (Ag) Max. 0.05 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 0.5 ppb
Barium (Ba) Max. 0.1 ppb
Beryllium (Be) Max. 0.1 ppb
Bismuth (Bi) Max. 0.05 ppb
Calcium (Ca) Max. 5 ppb
Cadmium (Cd) Max. 0.05 ppb
Cerium (Ce) Max. 0.05 ppb
Cobalt (Co) Max. 0.1 ppb
Cesium (Cs) Max. 0.05 ppb
Copper (Cu) Max. 0.5 ppb
Dysprosium (Dy) Max. 0.05 ppb
Erbium (Er) Max. 0.05 ppb
Europium (Eu) Max. 0.05 ppb
Iron (Fe) Max. 1 ppb
Gallium (Ga) Max. 0.05 ppb
Gadolinium (Gd) Max. 0.05 ppb
Holmium (Ho) Max. 0.05 ppb
Indium (In) Max. 0.05 ppb
Potassium (K) Max. 5 ppb
Lanthanum (La) Max. 0.05 ppb
Lithium (Li) Max. 0.05 ppb
Lutetium (Lu) Max. 0.05 ppb
Magnesium (Mg) Max. 0.5 ppb
Manganese (Mn) Max. 0.1 ppb
Molybdenum (Mo) Max. 0.1 ppb
Sodium (Na) Max. 10 ppb
Niobium (nb) Max. 0.05 ppb
Neodymium (nd) Max. 0.05 ppb
Nickel (Ni) Max. 0.1 ppb
Lead (Pb) Max. 0.2 ppb
Palladium (Pd) Max. 0.5 ppb
Praseodymium (Pr) Max. 0.05 ppb
Platinum (Pt) Max. 0.1 ppb
Rubidium (Rb) Max. 0.1 ppb
Rhodium (Rh) Max. 0.1 ppb
Antimony (Sb) Max. 0.1 ppb
Samarium (Sm) Max. 0.05 ppb
Tin (Sn) Max. 0.1 ppb
Strontium (Sr) Max. 0.1 ppb
Terbium (Tb) Max. 0.05 ppb
Thorium (Th) Max. 0.1 ppb
Titanium (Ti) Max. 0.5 ppb
Thallium (Tl) Max. 0.05 ppb
Thulium (Tm) Max. 0.05 ppb
Uran (U) Max. 0.05 ppb
Vanadium (V) Max. 0.2 ppb
Yttrium (Y) Max. 0.05 ppb
Ytterbium (Yb) Max. 0.05 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 2 ppb

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR