Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12386.FF 12386.FI
12386.FFEA 99.8 GBP
12386.FF 12386.FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Erbium

Purity: 99.9% CAS No.: 7440-52-0 Physical Form: ≈1.4g/25x25mm Odour: Odourless MDL No.: MFCD00010987

Formula: Er
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR