Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

12373.18 12373.30
12373.18EA 34.8 GBP
12373.18 12373.30
Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS
Cadmium (II) chloride hemipentahydrate

CAS No.: 7790-78-5 Molecular Formula: CdCl2.2.5H2O Formula Weight: 228.36 (183.32anhy) Physical Form: Crystalline Refractive Index: 1.6513 Sensitivity: Hygroscopic Merck Reference: 14,1617 UN No.: 2570 MDL No.: MFCD00799071

Formula: CdCl₂·2.5H₂O
Melting Pt: 568 °C
Density: 2.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00799071
CAS Number: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 14,1617

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR