Chromium(III) oxide ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Chromic oxide

12285.22 12285.36
12285.22EA 39.8 GBP
12285.22 12285.36
Chromium(III) oxide ≥99% (metals basis)
Chromium (III) oxide

Purity: 99% CAS No.: 1308-38-9 Molecular Formula: Cr2O3 Formula Weight: 151.99 Physical Form: Powder Density: 5.22 Refractive Index: 2.551 Appearance: Pale to dark green Odour: Odourless Melting Point: 2435° Boiling Point: 3000° Merck Reference: 14,2234 MDL No.: MFCD00010949

Formula: Cr₂O₃
Boiling Pt: 3000 °C
Melting Pt: 2435 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR