Strontium acetate hemihydrate, Reagent Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Strontium Acetate Hemihydrate

12203.30 12203.A1
12203.30EA 53.2 GBP
12203.30 12203.A1
Strontium acetate hemihydrate, Reagent Grade
Strontium acetate hemihydrate

Converts to SrCO₃ on ignition
Soluble in water


CAS No.: 14692-29-6 Molecular Formula: Sr(OOCCH3)2.0.5H2O Formula Weight: 214.72 (205.71anhy) Physical Form: Crystalline Melting Point: 150° -0.5H2O Solubility: Soluble in water MDL No.: MFCD00198064

Formula: C₈H₁₄O₉Sr₂
MW: 429.43 g/mol
Melting Pt: 150 °C (-0.5H₂O)
Density: 2.099
Storage Temperature: Ambient
MDL Number: MFCD00198064
CAS Number: 14692-29-6
EINECS: 208-854-8
Merck Index: 14,8836

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR