PIG® Oil-Only mat pads and rolls
Supplier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MAT403 MAT401 MAT440 MAT415 MAT419 MAT423 MAT424 MAT425 MAT460 MAT461 MAT462
121-0532EA 141 GBP
121-0532 121-0100 121-0106 121-0101 121-0102 121-0103 121-0104 121-0105 121-0107 121-0108 121-0109
PIG® Oil-Only mat pads and rolls
Spill Control Materials
Tough enough for rough handling, guaranteed. Ship your haz waste in our UN rated overpacks to stay compliant with regs.
  • Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water
More Product Information