PIG® universal mat rolls
Supplier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MAT230 MAT411 MAT283 MAT220 MAT202-01 MAT137 MAT140 MAT153 MAT225 MAT226 MAT245 MAT246 MAT263 MAT185
121-0522EA 293 GBP
121-0522 121-0533 121-0098 121-0090 121-0086 121-0080 121-0082 121-0083 121-0091 121-0092 121-0095 121-0096 121-0097 121-0085
PIG® universal mat rolls
Spill Control Materials
Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated.
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job longer
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
  • Can be wrung out and
More Product Information