Chemical classifier strips, Spilfyter®
Supplier: NPS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
570001 570010 570050
121-0024EA 94 GBP
121-0024 121-0039 121-0040
Chemical classifier strips, Spilfyter®
Environmental Test Kits
Strips simultaneously test for acid/base, oxidiser, fluoride, petroleum product/organic solvent, and iodine/bromine/chlorine risks.
  • Identify unknown spilt liquids and determine proper treatment in seconds
  • Chemical classifier chart determines possible presence of types according to strips colour changes
Identify unknown spilt liquids and determine proper treatment in seconds
Chemical classifier chart determines possible presence of types according to strips colour changes
Delivery information: Chemical classifier station includes 20 chemical classifier strips, 1 chemical classifier chart, 2 bobbins of polypropylene thread, and 1 weather-resistant plastic carry case. Chemical classifier kit includes 6 chemical classifier strips, 1 chemical classifier chart, and 1 bobbin of polypropylene thread. Additional strips and charts are available separately.