Safety canisters
Supplier: ROETZMEIER SICHERH BEHAELTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05KD 05KK 05KZ 10KD 10KK 10KZ 20KDL 20KK 20KZ
119-0172EA 370 GBP
119-0172 119-2045 119-2180 119-0116 119-2046 119-2181 119-2380 119-0121 119-2182
Safety canisters
Safety Cans
Stainless steel.
  • In-plant use of flammable liquids, such as solvents
  • Explosion-proof due to flame traps (removable) and pressure control valves
  • Taps and metering devices are self-closing
  • High quality, chemically resistant stainless steel (0,8 mm thick) with PTFE gaskets
  • Easy handling due to ergonomic design
  • Models with self-closing taps are equipped with a separate ventilation screw and are suitable for horizontal storage and decanting
Canister shape maximises the usage of limited storage space, e.g. in safety cabinets.