Safety can, oily waste
Supplier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
09200 09500 09100 09501 09101 09300 09301 09700 09701 09710
119-0245EA 82.8 GBP
119-0245 119-0249 119-0242 119-0250 119-0243 119-0246 119-0247 119-0252 119-0253 119-0255
Safety can, oily waste
Safety Cans
These oily waste cans with galvanised steel body construction with durable powder-coat finish are essential when using solvent soaked cloths and wiping rags. Rags and cloths soaked with thinners, linseed oil, combustible adhesives and other flammable liquids present a serious fire risk when discarded improperly.
  • Available in several sizes and various colours
  • Lid opens no more than 60° and stays closed when not in use which isolates the contents from fire sources and limits oxygen, virtually eliminating a risk of spontaneous combustion
Compliance withMore Product Information