Sharp disposal containers for hazardous waste, Sharpsafe®
Supplier: Hospidex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SHA-41005430 SHA-41005420 SHA-41152420 SHA-41802420 SHA-41172440 SHA-41455430 SHA-41602430 SHA-41202431 SHA-41802431 SHA-41202421
119-0078EA 425 GBP
119-0078 119-0182 119-0185 HUARSHA-41802420 HUARSHA-41172440 119-0191 119-0192 HUARSHA-41202431 HUARSHA-41802431 HUARSHA-41202421
Sharp disposal containers for hazardous waste, Sharpsafe®
Bins Sharps Containers
PP, yellow. Disposable plastic sharps container.
Orange lids – Sharps for incineration or alternative treatment. Marked 'Fully Discharged Sharps' the inscription and 'tick' indicate this coloured lid container is for use with sharps not containing prescription only medicines or 'POM's.
Yellow lids – Infectious sharps for incineration only. Marked with 'Medicinal Sharps', the inscription and 'tick' indicates this coloured lid container will be for general use, including sharps containing medicinal residue.
Purple lids – Marked with 'Cyto Sharps', the inscription and 'tick' indicateMore Product Information