Sodium tetrachloropalladate(II) hydrate (30% Pd) ≥99.95% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

11886.04 11886.09 11886.18
11886.04EA 219 GBP
11886.04 11886.09 11886.18
Sodium tetrachloropalladate(II) hydrate (30% Pd) ≥99.95% (metals basis)
Sodium tetrachloropalladate(II) hydrate

Purity: 99.95% CAS No.: 13820-53-6 Molecular Formula: Cl4Na2Pd.xH2O (x≈3) Formula Weight: 294.20(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red-brown MDL No.: MFCD00003487

Formula: Cl₄Na₂Pd·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003487
CAS Number: 13820-53-6
EINECS: 237-502-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR