Bathophenanthroline, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)

M-Clarity™ quality level = MQ100

11880-1G-F 11880-5G-F
11880-1G-FEA 114 GBP
11880-1G-F 11880-5G-F
Bathophenanthroline, Supelco®
Bathophenanthroline
Formula: C₂₄H₁₆N₂
Boiling Pt: 508 °C (1013 hPa)
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS Number: 1662-01-7
EINECS: 216-767-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb699d3517368ad7b525efa816297cc58ac8675&vid=5abaea183ad59c2796879d539bf80c751c673354&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR