Manganese(II) oxide 99% (76.0 - 78.0% Mn)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

11870.36 11870.A3 11870.F7
11870.36EA 32.9 GBP
11870.36 11870.A3 11870.F7
Manganese(II) oxide 99% (76.0 - 78.0% Mn)
Manganese (II) oxide

Purity: 99% CAS No.: 1344-43-0 Molecular Formula: MnO Formula Weight: 70.94 Physical Form: +200 Mesh Powder Density: 5.0 Appearance: Green Odour: Odourless MDL No.: MFCD00016226

Formula: MnO
Melting Pt: 1840 °C
Density: 5.0
Storage Temperature: Ambient
MDL Number: MFCD00016226
CAS Number: 1344-43-0
EINECS: 215-695-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR