Tungsten disulphide ≥99.8% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11829.18 11829.30
11829.18EA 63.1 GBP
11829.18 11829.30
Tungsten disulphide ≥99.8% (metals basis)
Tungsten disulphide

Purity: 99.8% CAS No.: 12138-09-9 Molecular Formula: WS2 Formula Weight: 247.98 Physical Form: Powder Density: 7.5 Appearance: Black Melting Point: 1250° dec. MDL No.: MFCD00011467

Formula: S₂W
Melting Pt: 1250 °C (dec.)
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00011467
CAS Number: 12138-09-9
EINECS: 235-243-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR