VWR®, Fire Blankets
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
118-1105EA 61.9 GBP
118-1105 118-1106 118-1107
VWR®, Fire Blankets
First Aid Supplies Fire and Emergency Blankets
Made from a composite material including two plies of glass fibre matting and one layer of flame-retardant foil. For extinguishing and smothering relatively small fires. Environmentally friendly, these blankets contain no asbestos.
  • Heat resistant up to 550 °C
  • Safe, dust-free storage in a covered plastic box labelled in three languages (German, English, French)
  • Can be wall mounted
  • Unfolds quickly with two ripcords
BS EN 1869
Certifications: BS EN 1869