Eyewash solutions in portable case
Supplier: ORKLA CARE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
190400
118-0253EA 41.9 GBP
118-0253
Eyewash solutions in portable case
First Aid Supplies Eyewash Stations and Eyewash Bottles
This plastic, case with transparent lid and handle, contains five bottles of Cederroth eyewash solution. The solution is intended for washing away particles and dangerous liquids, like alkali or acids, from an eye in an emergency situation in the workplace.
  • Portable case is ideal for mobile workplaces and for continuous irrigation during transport to the hospital
  • Buffered eyewash has a neutralising effect on splashes from alkalis and acids
  • Box made of green polystyrene, lid made of transparent ABS
  • Case can be wall mounted by using the wall bracket