Eyewash kit, Evolution Plus
Supplier: SAFETY FIRST AID GROUP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E459T
118-0117EA 107 GBP
118-0117
Eyewash kit, Evolution Plus
First Aid Supplies Eyewash Stations and Eyewash Bottles
This portable eyewash kit includes two HypaClens sterile eyewash bottles (500 ml each) and two HypaCover sterile eye dressings.
  • Emergency eye wash kit for any location
  • Self-supporting workbench allows kit to be opened and worked with on the bracket
  • Durable hinged case includes bottle stabilisers to hold contents firmly in place
  • Supplied with a quick release wall fixing bracket and kit identification label
  • 0,9% sterile saline solution is ideal for irrigating eyes and general wound washing applications