Palladium 5% on barium carbonate, powder hydrogenation catalyst, reduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

11721.06 11721.14 11721.22
11721.06EA 0 GBP
11721.06 11721.14 11721.22
Palladium 5% on barium carbonate, powder hydrogenation catalyst, reduced
Palladium

Odour: Odourless UN No.: 1564 MDL No.: MFCD03427451

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 1564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR