Magbeads FastDNA kit, 192 preparations, MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
116575192
116575192.EA 932 GBP
116575192.
Magbeads FastDNA kit, 192 preparations, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MagBeads FastDNA® Kit is intended for rapid extraction of DNA from tissue, cells, blood, saliva, swabs, blood spots, semen, and other clinical samples. DNA can be used directly for PCR, quantitative PCR, Southern Blot, detection of viral DNA and so on.
  • High quality DNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals
MagBeads FastDNA® Kit is based on the purification method of high binding magnetic particles. The sample is lysed and digested. RNA is released into the lysate. After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts. Finally the RNA was eluted with elution buffer.