Revolve™ Sustainable Sterile Pre-Wetted Cleanroom Wipes
Supplier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
STX1704P STX1709P STX1712P
115-2708EA 295 GBP
115-2708 115-2711 115-2713
Revolve™ Sustainable Sterile Pre-Wetted Cleanroom Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment and parts
  • Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol,
More Product Information