Cleanroom wipes, ReFIBE™
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RFHS-1212 RFHS-99 RFHS-1212IR RFHS-99IR
115-2510EA 715 GBP
115-2510 115-2511 115-3074 115-2679
Cleanroom wipes, ReFIBE™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Recycled 100% knitted polyester cleanroom wipes offer a sustainable solution for cleanroom environments, without jeopardising quality or performance. Soft textured wipes clean surfaces without scratching, while offering low levels of particles and fibres, abrasion and chemical resistance, and good solvent compatibility.
  • 100% knitted laundered polyester
  • Laser cut sealed edges seals fibres at wipe's edge to reduce shedding
  • Suitable for use in high grade cleanrooms
  • Manufactured from 100% recycled plastic
Each wipe is constructed from 100% post-consumerMore Product Information