Cleanroom wipes, sterile, Polynit Heatseal Low Endotoxin
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LWLE0001 LWLE0002
115-2438EA 647 GBP
115-2438 115-2448
Cleanroom wipes, sterile, Polynit Heatseal Low Endotoxin
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
These wipes are manufactured from high quality 100% knitted polyester fabric with sealed edges. The polyester fabric offers the widest range of solvent compatibility. Ideal for use in product contact areas.
  • Good solvent sorbency, abrasion and chemical resistance
  • Extremely low in particles and fibres
  • Eendotoxin level of <1 EU/wipe
  • Validated sterile to 10⁻⁶ Sterility Assurance Level
  • Small quantity of wipes per package can be used during one cleaning session, eliminating waste
Suitable for use in ISO Class 5 (Grade A/B) cleanroom environments.