Cleaning wipe, microfiber
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K-1561
115-2151EA 33 GBP
115-2151
Cleaning wipe, microfiber
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
These microfibre wipes are ideal for the smear-free cleaning of glass, chrome, plastics and optical lenses.
  • For cleaning without detergents
  • Can be used either wet or dry
  • Washable up to 90 °C
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904