Wipe rolls, WYPALL* L40
Supplier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7452
115-2051EA 322 GBP
115-2051
Wipe rolls, WYPALL* L40
Wipes Disposable Wipes and Towels Wipes for General Applications
These single ply wipes are made from AIRFLEX* material, a specialised fabric with a perfect balance of softness and strength, that is also thick and absorbent. Ideal for industrial tasks such as cleaning, process line wiping, soaking up large spills.
  • Made with double re-creped (DRC) base sheet technology
  • Large, perforated rolls for use in busy areas
  • Also suitable for use in healthcare for wiping patients’ hands and face
  • Can be used with a dispensing system to help control use and reduce wastage
Matching dispensers: 115-2038, 115-0045 and 115-1533.