Sterile chemotherapy prep bench mat, BioClean ChemPrep™
Supplier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S-BCPM-4025B S-BCPM-5640B
115-1880EA 686 GBP
115-1880 115-1938
Sterile chemotherapy prep bench mat, BioClean ChemPrep™
Surface Protectors Bench Protectors
The sterile chemotherapy prep mat from BioClean is highly absorbent.
  • Three-layer construction
  • Mesh upper layer for even distribution, highly absorbent middle layer, impermeable bottom layer
  • Sterilised by gamma radiation, low-linting, latex-free
Certifications: ISO 9001:2008, EU GMP Grade A compatible
Caution: To be stored in a dry, cool place (up to 40 °C) away from direct sunlight and fluorescent light.