Disinfectant wipes
Supplier: Synergy Health
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
81103
115-0709EA 73.4 GBP
115-0709
Disinfectant wipes
Wipes Disposable Wipes and Towels Disinfectant Wipes
Non-woven wipes for disinfection of clinical hard surfaces. Store away from heat and do not expose to naked flame.
  • Impregnated with 70% v/v solution of isopropyl alcohol BP
Packaging: Supplied in dispensing tub.
Caution: Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904