Cleanroom wipes, multipurpose, Amplitude™ EcoCloth™
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AMEC0003 AMEC0004 AMEC0005 AMEC0001 AMEC0002
115-0570EA 32.4 GBP
115-0570 115-0571 115-0572 115-0569 115-0608
Cleanroom wipes, multipurpose, Amplitude™ EcoCloth™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Absorbent, non woven cellulose/polyester wipes, feature enhanced sorbency while being economical due to their light weight.
  • Lightweight, highly sorbent wipe, designed for non abrasive applications
  • Strong and durable hydroentangled fabric with good solvent compatibility
  • Low levels of particles and extractables
Recommended for general purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical device and printed circuit board industries. Ideal for wiping products prior to introduction into a cleanroom, or wiping and absorbing spills.More Product Information