Flat mop covers TechniCloth®
Supplier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TX627C TX630
115-0480EA 123 GBP
115-0480 TEXWTX630
Flat mop covers TechniCloth®
Mops
Mop covers (ISO Class 3 - 8) for cleaning and decontaminating floors.
  • Made without chemical binders or additives
  • Non-woven, economical polyester/cellulose for cleaning and removing residues
  • Chemically resistant, withstand exposure to common cleaning and disinfecting solutions