Cleanroom wipes, Polynit Heatseal
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PNHS-1212 PNHS-44 PNHS-99 PNHS-99B/150 PNHS-66 LWPS0030 PNHS-99B PNHS-44B LWPN0020
115-0375EA 73.6 GBP
115-0375 115-0376 115-0268 CONTPNHS-99B/150 115-2148 CONTLWPS0030 CONTPNHS-99B CONTPNHS-44B CONTLWPN0020
Cleanroom wipes, Polynit Heatseal
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment. Made from 100% textured polyester no-run knit.
  • Good solvent sorbency, abrasion and chemical resistance
  • Extremely low in particles and fibres
  • Laser cutting seals fibres for non abrasive edges
Suitable for use in Class 1, ISO 3, M1.5 cleanrooms
Colour: White.