Cleanroom wipes, sterile, Gamma Wipe® 300
Supplier: BERKSHIRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GW300.ST.25
115-0261EA 2110 GBP
115-0261
Cleanroom wipes, sterile, Gamma Wipe® 300
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
This sterile wipe is made of a continuous-filament 100% polyester knit. It is knitted in tube form and sealed at the two open edges for optimal cleanliness. Laundered in an ISO Class 4 (Class 10) cleanroom.
  • Two ply construction provides tremendous absorbency and durability
  • Wipe is abrasion and solvent-resistant
  • Excellent wet strength, will withstand sanitisers and disinfectants such as bleaches, phenols
  • Certificate of Sterility is included with each case
Suitable for use in Class 1, ISO 3, M1.5 environments.
Sterilised by gamma radiation to aMore Product Information